Orders LinkContactUsLinkComponent
DNA markers
DNA markers
DNA marker, 1 - 10 kb, 2× 250 μl, 200
Katalog # LONZ50471
Leverandør:   LONZA
DNA markers
Katalog # LONZ50471
Leverandør:  LONZA
CAS nummer:  
DNA markers
DNA markers
DNA marker, 1 - 10 kb, 2× 250 μl, 200
CAS nummer:  
Price
4.730,00 kr
Log ind for tilgængelighed
1 * 200 Items
Product Details & Documents
DNA marker is for rapid size estimation of PCR products and restriction fragments. DNA can be visualised by ethidium bromide staining or kinased with radiolabelled ³²P for detection by autoradiography.
Specifikationer
Specifikationer
  • Pakn:
    200
  • Beskrivelse:
    DNA marker, 1 - 10 kb, 2× 250 μl