DNA markers
DNA marker, 1 - 10 kb, 2× 250 μl, 200
Katalog # LONZ50471
Leverandør: LONZA
Gå til:
Product Details & Documents
DNA marker is for rapid size estimation of PCR products and restriction fragments. DNA can be visualised by ethidium bromide staining or kinased with radiolabelled ³²P for detection by autoradiography.
Specifikationer
Specifikationer
- Pakn:200
- Beskrivelse:DNA marker, 1 - 10 kb, 2× 250 μl