Orders LinkContactUsLinkComponent
DNA markers
DNA markers
DNA marker, 50 - 1000 bp, 250 μl, 50
Katalog # LONZ50461
Leverandør:   LONZA
DNA markers
Katalog # LONZ50461
Leverandør:  LONZA
CAS nummer:  
DNA markers
DNA markers
DNA marker, 50 - 1000 bp, 250 μl, 50
CAS nummer:  
Price
4.040,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents
DNA marker is for rapid size estimation of PCR products and restriction fragments. DNA can be visualised by ethidium bromide staining or kinased with radiolabelled ³²P for detection by autoradiography.
Specifikationer
Specifikationer
  • Pakn:
    50
  • Beskrivelse:
    DNA marker, 50 - 1000 bp, 250 μl