Agarose, SeaKem® HEEO
Agarose, SeaKem® HEEO
Supplier: LONZA
Jump to:
Product Details & Documents
SeaKem® HEEO Agarose is a very high EEO agarose that is useful in applications requiring significant cathodal migration, such as IEP of IgG and IgM. It can also be used to blend with a lower EEO agarose to achieve a specific EEO value.
Documents
Certificates
Search by Lot NumberSpecifications
- Beskrivelse:Agarose, SeaKem® HEEO
- Seq:02