Orders LinkContactUsLinkComponent
Agarose, SeaKem® HE
Agarose, SeaKem® HE
Agarose, SeaKem® HE
Katalog # 733-1556
Leverandør:   LONZA
Agarose, SeaKem® HE
Katalog # 733-1556
Leverandør:  LONZA
Agarose, SeaKem® HE
Agarose, SeaKem® HE
Agarose, SeaKem® HE
Price
1.870,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
SeaKem® HE agarose is a high EEO agarose that provides enhanced resolution in immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CIEP), and serum protein electrophoresis.
Specifikationer
  • Beskrivelse:
    Agarose, SeaKem® HE
  • Seq:
    01