Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
SeaKem® ME Agarose

SeaKem® ME Agarose

Glass bottle for solids, 25 g
Katalog # 733-1554
Leverandør:   LONZA
SeaKem® ME Agarose
Katalog # 733-1554
Leverandør:  LONZA
CAS nummer:  9012-36-6
SeaKem® ME Agarose
SeaKem® ME Agarose

Glass bottle for solids, 25 g

CAS nummer:  9012-36-6
Price
1.800,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
  • CAS nummer:9012-36-6
  • MDL nummer:MFCD00081294
  • Smeltende Pt:60…90 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-731-8

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.