Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumthiosulfat, anhydrous 99%

Natriumthiosulfat, anhydrous 99%

Glass bottle for solids, 500 g
Catalog # A17629.36
Natriumthiosulfat, anhydrous 99%
Catalog # A17629.36
CAS Number:  7772-98-7
Natriumthiosulfat, anhydrous 99%
Natriumthiosulfat, anhydrous 99%

Glass bottle for solids, 500 g

CAS Number:  7772-98-7
Price
370,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Odour: Odourless Merck Reference: 14,8694 MDL No.: MFCD00003499

Synonyms:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS Number:7772-98-7
  • Formula:Na₂S₂O₃
  • MDL Number:MFCD00003499
  • Molecular Weight:158.11 g/mol
  • Merck Index:14,8694
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:48 °C
  • Density:1,667
  • Storage Temperature:Stue temperatur
  • EINECS:231-867-5
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifications