Orders LinkContactUsLinkComponent
Natriumthiosulfat, anhydrous 99%
Glass bottle for solids, 2.500 g
  A17629.0E
Natriumthiosulfat, anhydrous 99%
  A17629.0E
 :  7772-98-7
Natriumthiosulfat, anhydrous 99%
Natriumthiosulfat, anhydrous 99%
Glass bottle for solids, 2.500 g
 :  7772-98-7
1.410,00 kr
1 * 2.500 g
Purity: 99% CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Odour: Odourless Merck Reference: 14,8694 MDL No.: MFCD00003499
 :  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  •  :7772-98-7
  •  :Na₂S₂O₃
  •  :MFCD00003499
  •  :158.11 g/mol
  •  :14,8694
  •  :100 °C (1013 hPa)
  •  :48 °C
  •  :1,667
  •  :Stue temperatur
  •  :231-867-5
  • Pakning:
    Glass bottle for solids
  • Pakn:
    2.500 g