Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Batterier

Batterier

Rechargable NIMH Power ACCU™, Mignon AA/HR6, 1,2 V, 2100 mAh
Catalog # VART56706101402
Batterier
Catalog # VART56706101402
Batterier
Batterier

Rechargable NIMH Power ACCU™, Mignon AA/HR6, 1,2 V, 2100 mAh

Price
168,00 kr
Log in for availability
1 * 2 Items
Product Details & Documents

Høj kapacitet med lange driftstider pr. opladning. Kræver ikke en speciel oplader.

  • Kan genoplades op til 1000 gange
Specifications
  • Model:
    Mignon AA/HR6
  • Spænding:
    1,2 V
  • Kapacitet:
    2100 mAh
  • Beskrivelse:
    Rechargable NIMH Power ACCU™
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.