Microplates, 96-well
96-brønds mikroplade, F-bund (skorstensbrønd), MICROLON® 200, 25 - 340 µl, 392 µl
Katalog # 738-0017
Leverandør: Greiner Bio-One
Gå til:
Product Details & Documents
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
- Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Documents
Specifikationer
- Beskrivelse:96-brønds mikroplade, F-bund (skorstensbrønd), MICROLON® 200
- Seq:07
- Arbejdsvolumen:25 - 340 µl
- Total volumen:392 µl
- Cat. No.:738-0017
- Miljøcertificering:ISO 14001 (241002); ISO 50001 (241002)
- Brøndprofil:F-bund (skorstensbrønd)
- Environmentally Preferable:Lav fremstillingspåvirkning
- Driftsvolumen:25 - 340