Thiamin hydrochlorid
Glass bottle for solids, 100 g
Katalog # SERA36020-0100
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
- CAS nummer:67-03-8
- Formel:C₁₂H₁₈Cl₂N₄OS
- MDL nummer:MFCD00012780
- Molecular Weight:337.27 g/mol
- Merck Index:13,09366
- Smeltende Pt:252 °C
- Tæthed:1,401 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-641-8
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



