Labels, hazard warning, electricity
Hazard labels, Polyester, Sort på gul, 100×87 mm, 3 labels/card
Katalog # 116-0757
Leverandør: Brady
Gå til:
Product Details & Documents
These electricity hazard warning labels are made of polyester and are used for prevention of accidents.
- Self-adhesive
- Clear identification of voltage, phases, grounding and insulation
Specifikationer
- Farve:Sort på gul
- Beskrivelse:Hazard labels
- Dimensioner:100×87 mm
- Materiale:Polyester
- Seq:02
- Pakket:3 labels/card