Orders LinkContactUsLinkComponent
Adapters, cone/screw thread, Quickfit®
Adapters, cone/screw thread, Quickfit®
14/23 NS, 18 mm, 7,0 - 8,5 mm
Katalog # 201-0696
Leverandør:   SciLabware
Adapters, cone/screw thread, Quickfit®
Katalog # 201-0696
Leverandør:  SciLabware
Adapters, cone/screw thread, Quickfit®
Adapters, cone/screw thread, Quickfit®
14/23 NS, 18 mm, 7,0 - 8,5 mm
Price
681,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer. Ideal for securing thermometers, gas inlet tubes etc.
Specifikationer
  • Konusstr.:
    14/23 NS
  • Hul Ø:
    7,0 - 8,5 mm
  • Gevind:
    18 mm
  • Seq:
    02