Wine refractometers, Master series
Wine refractometer, MASTER-BAUMÉ, 0 − 21 °Bé ; 0 − 40 °Bx, 0,2 °Bé ; 0,2 °Bx
Katalog # ATAG2594
Leverandør: Atago
Gå til:
Product Details & Documents
Compact and portable instruments for indoor or outdoor use.
They can either fit in a pocket or be hanged by a strap.
Specifikationer
- Model:MASTER-BAUMÉ
- Område:0 − 21 °Bé ; 0 − 40 °Bx
- Opløsning:0,2 °Bé ; 0,2 °Bx
- Beskrivelse:Wine refractometer
- Cat. No.:ATAG2594
- Vægt:160 g