Orders LinkContactUsLinkComponent
Wine refractometers, Master series
Wine refractometers, Master series
Wine refractometer, MASTER-BAUMÉ, 0 − 21 °Bé ; 0 − 40 °Bx, 0,2 °Bé ; 0,2 °Bx
Katalog # ATAG2594
Leverandør:   Atago
Wine refractometers, Master series
Katalog # ATAG2594
Leverandør:  Atago
Wine refractometers, Master series
Wine refractometers, Master series
Wine refractometer, MASTER-BAUMÉ, 0 − 21 °Bé ; 0 − 40 °Bx, 0,2 °Bé ; 0,2 °Bx
Price
1.570,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Compact and portable instruments for indoor or outdoor use.
They can either fit in a pocket or be hanged by a strap.
Specifikationer
  • Model:
    MASTER-BAUMÉ
  • Område:
    0 − 21 °Bé ; 0 − 40 °Bx
  • Opløsning:
    0,2 °Bé ; 0,2 °Bx
  • Beskrivelse:
    Wine refractometer
  • Cat. No.:
    ATAG2594
  • Vægt:
    160 g