Wine refractometers, Master series
Wine refractometer, MASTER-GOE, 30 − 170 °Oe ; 0 − 40 °Bx, 1 °Oe ; 0,2 °Bx
ATAG2592
: Atago
Compact and portable instruments for indoor or outdoor use.
They can either fit in a pocket or be hanged by a strap.
- Model:MASTER-GOE
- Område:30 − 170 °Oe ; 0 − 40 °Bx
- Opløsning:1 °Oe ; 0,2 °Bx
- Beskrivelse:Wine refractometer
- Cat. No.:ATAG2592
- Vægt:160 g