Orders LinkContactUsLinkComponent
Wine refractometers, Master series
Wine refractometer, MASTER-GOE, 30 − 170 °Oe ; 0 − 40 °Bx, 1 °Oe ; 0,2 °Bx
  ATAG2592
 :   Atago
Wine refractometers, Master series
  ATAG2592
 :  Atago
Wine refractometers, Master series
Wine refractometers, Master series
Wine refractometer, MASTER-GOE, 30 − 170 °Oe ; 0 − 40 °Bx, 1 °Oe ; 0,2 °Bx
1.570,00 kr
1 * 1 Items
Compact and portable instruments for indoor or outdoor use.
They can either fit in a pocket or be hanged by a strap.
  • Model:
    MASTER-GOE
  • Område:
    30 − 170 °Oe ; 0 − 40 °Bx
  • Opløsning:
    1 °Oe ; 0,2 °Bx
  • Beskrivelse:
    Wine refractometer
  • Cat. No.:
    ATAG2592
  • Vægt:
    160 g