Accessories for SCAT Level Controls
Dual lock (releasable pressure) closure for disc sensors, 20×20 mm
Katalog # 554-2714
Leverandør: S.C.A.T.
Gå til:
Product Details & Documents
Necessary and helpful accessories for use with SCAT electronic level controls.
- Switching cables
- Disk sensors
- Signal cables
- Power supply units
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifikationer
- Beskrivelse:Dual lock (releasable pressure) closure for disc sensors, 20×20 mm
- Seq:16
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



