Orders LinkContactUsLinkComponent
Scintillation vials, PET, WHEATON®
Scintillation vials, PET, WHEATON®
20,00 ml, 27×61* mm, Yes, Urea, Metal foil
Katalog # 548-2046
Leverandør:   DWK Life Sciences
Scintillation vials, PET, WHEATON®
Katalog # 548-2046
Leverandør:  DWK Life Sciences
Scintillation vials, PET, WHEATON®
Scintillation vials, PET, WHEATON®
20,00 ml, 27×61* mm, Yes, Urea, Metal foil
Price
3.740,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Manufactured from clear polyethylene terephthalate (PET) these vials have advantages over glass counting vials of low background count, economy and virtual unbreakability.
  • Clarity of glass
  • Can be safely incinerated
  • Lightweight and virtually unbreakable
* With cap on
Specifikationer
  • Kasket inkluderet:
    Yes
  • Hættemateriale:
    Urea
  • Diameter×H:
    27×61* mm
  • Septum:
    Metal foil
  • Kapacitet:
    20,00 ml