Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Supelco® Organic Reference Standard, Cyclohexene

Glass bottle, 10 mL
Katalog # 44028-10ML-F
Leverandør:   Merck
Supelco® Organic Reference Standard, Cyclohexene
Katalog # 44028-10ML-F
Leverandør:  Merck
CAS nummer:  
Supelco® Organic Reference Standard, Cyclohexene
Supelco® Organic Reference Standard, Cyclohexene

Glass bottle, 10 mL

CAS nummer:  
Price
Spørg for pris
Log ind for tilgængelighed
1 * 10 mL
Product Details & Documents
  • GHS signal: GHS02, GHS06, GHS08

 : P210 - P233 - P240 - P273 - P301 + P310 - P331

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    10 mL

Specifikationer