EDTA ≥99.0% for complexometry, Fluka™
Plastic bottle, 1 kg
Leverandør: Honeywell Chemicals
EDTA ≥99.0% for complexometry, Fluka™
Katalog # 03610-1KG
Leverandør: Honeywell Chemicals
CAS nummer: 60-00-4
Gå til:
Product Details & Documents
Synonymer: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- CAS nummer:60-00-4
- Formel:C₁₀H₁₆N₂O₈
- MDL nummer:MFCD00003541
- Molecular Weight:292.25 g/mol
- Merck Index:13,03546
- Kogende Pt:614 °C (1013 hPa)
- Smeltende Pt:240 °C
- Tæthed:0,86 g/cm³ (20 °C)
- Flash Pt:≥ 100 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-449-4
- NÅ:01-2119486399-18
Documents
sds_03610
SDS
Specifikationer
Specifikationstestresultater
- Assay:99.0 - 101.0 %
- Identity (IR):complying
- Residue on ignition:Max. 0.1 %
- Colour of the solution:complying
- Clearness of the solution:complying
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 50 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 500 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Chloride (Cl):Max. 50 ppm
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



