Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ethylbenzen, Supelco®

Ethylbenzen, Supelco®

Glass bottle, 10 mL
Catalog # 03079-10ML
Flammable
Irritant
Supplier:   Merck
Ethylbenzen, Supelco®
Catalog # 03079-10ML
Supplier:  Merck
CAS Number:  100-41-4
Ethylbenzen, Supelco®
Ethylbenzen, Supelco®

Glass bottle, 10 mL

CAS Number:  100-41-4
Price
905,00 kr
Log in for availability
1 * 10 mL
Product Details & Documents
Synonyms:  Phenylethane
  • CAS Number:100-41-4
  • Formula:C₈H₁₀
  • MDL Number:MFCD00011647
  • Molecular Weight:106.17 g/mol
  • Merck Index:13,03801
  • Boiling Pt:136,2 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:0,867 g/cm³ (20 °C)
  • Flash Pt:15 °C
  • Storage Temperature:Stue temperatur
  • UN:1175
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:202-849-4

Documents

Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    10 mL

Specifications