Supelco® Organic Reference Standard, Methanol
Methanol, Analytical standard, 10 mL
: Merck
- GHS signal: GHS02, GHS06, GHS08
: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311
- Pakn:10 mL
- Standard:Analytical standard
- Beskrivelse:Methanol
- Densitet:0,791 g/ml at 25 °C (lit.)
- MDL No.:MFCD00004595
- Synonym(s):Methyl alcohol
- Formel:CH₃OH
- Cat. No.:82762-10ML-F
- M-Clarity quality level:MQ100
- CAS-nr:67-56-1
- Molekylvægt:32,04
- Kogepunkt:64.7 °C (lit.)
- Fareklassebeskrivelse:H225 - H301 + H311 + H331 - H370
- Flammepunkt:9,7 °C - Closed cup
- Opbevaring betingelser:2...30 °C
- Melting point:–98 °C (lit.)
- Shipping conditions:STD



