Orders LinkContactUsLinkComponent
Buffer solutions, according to DIN 19266, Fluka™
Buffer solutions, according to DIN 19266, Fluka™
10.012, ±.020, Bicarbonater, 25 °C, Plastic bottle, 1×500 ml, 500 mL
Katalog # 82454-500ML
Leverandør:   Honeywell Chemicals
Buffer solutions, according to DIN 19266, Fluka™
Katalog # 82454-500ML
Leverandør:  Honeywell Chemicals
CAS nummer:  
Buffer solutions, according to DIN 19266, Fluka™
10.012, ±.020, Bicarbonater, 25 °C, Plastic bottle, 1×500 ml, 500 mL
CAS nummer:  
Price
1.280,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Buffers with fungicide (except pH 10,012)
Certifications: According to DIN 19266.
Documents
Specifikationer
Specifikationer
  • Pakning:
    Plastic bottle
  • pH buffer type:
    Bicarbonater
  • Pakn:
    500 mL
  • Referencetemperatur:
    25 °C
  • pH nøjagtighed:
    ±.020
  • Pk Info:
    1×500 ml
  • pH-værdi:
    10.012