Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, 1-Decene

Supelco® Organic Reference Standard, 1-Decene

Glass bottle, 5 mL
Catalog # 30649-5ML
Supplier:   Merck
Supelco® Organic Reference Standard, 1-Decene
Catalog # 30649-5ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, 1-Decene
Supelco® Organic Reference Standard, 1-Decene

Glass bottle, 5 mL

CAS Number:  
Price
513,00 kr
Log in for availability
1 * 5 mL
Product Details & Documents
  • GHS signal: GHS02, GHS08, GHS09

Caution: P210 - P233 - P240 - P273 - P301 + P310 - P331

Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    5 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.