Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

n-Decan, Supelco®

Glass bottle, 5 mL
Katalog # 30540-5ML
Brandfarlig
Sundhed
Leverandør:   Merck
n-Decan, Supelco®
Katalog # 30540-5ML
Leverandør:  Merck
CAS nummer:  124-18-5
n-Decan, Supelco®
n-Decan, Supelco®

Glass bottle, 5 mL

CAS nummer:  124-18-5
Price
610,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents
  • CAS nummer:124-18-5
  • Formel:H₃C(CH₂)₈CH₃
  • MDL nummer:MFCD00008954
  • Molecular Weight:142.28 g/mol
  • Kogende Pt:174,1 °C (1013 hPa)
  • Smeltende Pt:–30 °C
  • Tæthed:0,73 g/cm³ (20 °C)
  • Flash Pt:46 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-686-4
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • FN:2247
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    5 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.