n-Decan, Supelco®
Glass bottle, 25 mL
Katalog # 30540-25ML
Leverandør: Merck
Gå til:
Product Details & Documents
- CAS nummer:124-18-5
- Formel:H₃C(CH₂)₈CH₃
- MDL nummer:MFCD00008954
- Molecular Weight:142.28 g/mol
- Kogende Pt:174,1 °C (1013 hPa)
- Smeltende Pt:–30 °C
- Tæthed:0,73 g/cm³ (20 °C)
- Flash Pt:46 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:204-686-4
- ADR:3,III
- :M-Clarity™ quality level = MQ100
- FN:2247
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:25 mL



