Cyclopentan, Supelco®
Glass bottle, 10 mL
Katalog # 29680-10ML
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Pentamethylene
- CAS nummer:287-92-3
- Formel:C₅H₁₀
- MDL nummer:MFCD00001356
- Molecular Weight:70.13 g/mol
- Merck Index:13,02769
- Kogende Pt:49 °C (1013 hPa)
- Smeltende Pt:–93,3 °C
- Tæthed:0,746 g/cm³ (20 °C)
- Flash Pt:–41 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1146
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- EINECS:206-016-6
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:10 mL