: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
- :60-33-3
- :C₁₈H₃₂O₂
- :MFCD00064241
- :280.45 g/mol
- :13,05527
- :360 °C (1013 hPa)
- :–5 °C
- :0,9 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- :Stue temperatur
- :200-470-9