Orders LinkContactUsLinkComponent
Linolsyre, Supelco®
Glass bottle, 5 mL
  62230-5ML-F
 :   Merck
Linolsyre, Supelco®
  62230-5ML-F
 :  Merck
 :  60-33-3
Linolsyre, Supelco®
Linolsyre, Supelco®
Glass bottle, 5 mL
 :  60-33-3
883,00 kr
1 * 5 mL
 :  (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
  •  :60-33-3
  •  :C₁₈H₃₂O₂
  •  :MFCD00064241
  •  :280.45 g/mol
  •  :13,05527
  •  :360 °C (1013 hPa)
  •  :–5 °C
  •  :0,9 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  •  :Stue temperatur
  •  :200-470-9
  • Pakning:
    Glass bottle
  • Pakn:
    5 mL