Cyclopentanon, Supelco®
Glass bottle, 50 mL
Katalog # 08299-50ML-F
Leverandør: Merck
Gå til:
Product Details & Documents
- CAS nummer:120-92-3
- Formel:C₅H₈O
- MDL nummer:MFCD00001409
- Molecular Weight:84.12 g/mol
- Merck Index:13,02771
- Kogende Pt:130 °C (1013 hPa)
- Smeltende Pt:–58 °C
- Tæthed:0,941 g/cm³ (20 °C)
- Flash Pt:31 °C
- Opbevaringstemperatur:Stue temperatur
- FN:2245
- ADR:3,III
- :M-Clarity™ quality level = MQ100
- EINECS:204-435-9
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:50 mL