Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(S)-(-)-Limonen, Supelco®

(S)-(-)-Limonen, Supelco®

Glass bottle, 5 mL
Katalog # 62128-5ML
Brandfarlig
Irriterende
Miljø
Leverandør:   Merck
(S)-(-)-Limonen, Supelco®
Katalog # 62128-5ML
Leverandør:  Merck
CAS nummer:  5989-54-8
(S)-(-)-Limonen, Supelco®
(S)-(-)-Limonen, Supelco®

Glass bottle, 5 mL

CAS nummer:  5989-54-8
Price
1.300,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents
Synonymer:  (S)-(-)-p-Mentha-1,8-dien, (S)-(-)-Dipenten, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexen, L(-)-Limonen
  • CAS nummer:5989-54-8
  • Formel:C₁₀H₁₆
  • MDL nummer:MFCD00001558
  • Molecular Weight:136.24 g/mol
  • Merck Index:13,05515
  • Kogende Pt:175 °C (1013 hPa)
  • Smeltende Pt:–74 °C
  • Tæthed:0,84 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:2052
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:227-815-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    5 mL

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.