Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Micro particles based on polystyrene, analytical standard, Supelco®

Micro particles based on polystyrene, analytical standard, Supelco®

15 µm, Aqueous suspension, 10% solids, std dev <0,2 µm, Analytical standard, 10 mL
Katalog # 74964-10ML-F
Leverandør:   Merck
Micro particles based on polystyrene, analytical standard, Supelco®
Katalog # 74964-10ML-F
Leverandør:  Merck
CAS nummer:  
Micro particles based on polystyrene, analytical standard, Supelco®

15 µm, Aqueous suspension, 10% solids, std dev <0,2 µm, Analytical standard, 10 mL

CAS nummer:  
Price
3.510,00 kr
Log ind for tilgængelighed
1 * 10 mL
Product Details & Documents

For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.

Specifikationer

Specifikationer

  • Partikelstørrelse:
    15 µm
  • Pakn:
    10 mL
  • Standard:
    Analytical standard
  • Beskrivelse:
    Aqueous suspension, 10% solids
  • Nøjagtighed:
    std dev <0,2 µm
  • Cat. No.:
    74964-10ML-F
  • M-Clarity quality level:
    MQ100

Specifikationer