(R)-(-)-Linalool, Supelco®
Glass bottle, 5 mL
Katalog # 74856-5ML-F
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: (-)-Linalool, (R)-3,7-Dimethylocta-1,6-dien-3-ol, (R)-(-)-3,7-Dimethyl-1,6-octadien-3-ol, L-Linalool
- CAS nummer:126-91-0
- Formel:C₁₀H₁₈O
- MDL nummer:MFCD00135469
- Molecular Weight:154.25 g/mol
- :M-Clarity™ quality level = MQ100
- EINECS:204-811-2
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:5 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



