(R)-(-)-Linalool, Supelco®
Glass bottle, 1 mL
Catalog # 74856-1ML-F
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: (-)-Linalool, (R)-3,7-Dimethylocta-1,6-dien-3-ol, (R)-(-)-3,7-Dimethyl-1,6-octadien-3-ol, L-Linalool
- CAS Number:126-91-0
- Formula:C₁₀H₁₈O
- MDL Number:MFCD00135469
- Molecular Weight:154.25 g/mol
- :M-Clarity™ quality level = MQ100
- EINECS:204-811-2
Specifications
Specifications
- Pakning:Glass bottle
- Pakn:1 mL



