n-Octan, Supelco®
74820-5ML
: 111-65-9
- :111-65-9
- :H₃C(CH₂)₆CH₃
- :MFCD00009556
- :114.23 g/mol
- :13,06782
- :126 °C (1013 hPa)
- :–57 °C
- :0,703 g/cm³ (20 °C)
- :8 °C
- :Stue temperatur
- :1262
- :3,II
- :M-Clarity™ quality level = MQ100
- :203-892-1
Pakning:
Glass bottle
Pakn:
5 mL