Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Micro particles based on polystyrene, analytical standard, Supelco®

Micro particles based on polystyrene, analytical standard, Supelco®

20 µm, Aqueous suspension, 10% solids, std dev <0,3 µm, Analytical standard, 5 mL
Katalog # 74491-5ML-F
Leverandør:   Merck
Micro particles based on polystyrene, analytical standard, Supelco®
Katalog # 74491-5ML-F
Leverandør:  Merck
CAS nummer:  
Micro particles based on polystyrene, analytical standard, Supelco®

20 µm, Aqueous suspension, 10% solids, std dev <0,3 µm, Analytical standard, 5 mL

CAS nummer:  
Price
2.270,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents

For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.

Specifikationer

Specifikationer

  • Partikelstørrelse:
    20 µm
  • Pakn:
    5 mL
  • Standard:
    Analytical standard
  • Beskrivelse:
    Aqueous suspension, 10% solids
  • Nøjagtighed:
    std dev <0,3 µm
  • Cat. No.:
    74491-5ML-F
  • M-Clarity quality level:
    MQ100

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.