Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Micro particles based on polystyrene, analytical standard, Supelco®

20 µm, Aqueous suspension, 10% solids, std dev <0,3 µm, Analytical standard, 10 mL
Catalog # 74491-10ML-F
Supplier:   Merck
Micro particles based on polystyrene, analytical standard, Supelco®
Catalog # 74491-10ML-F
Supplier:  Merck
CAS Number:  
Micro particles based on polystyrene, analytical standard, Supelco®

20 µm, Aqueous suspension, 10% solids, std dev <0,3 µm, Analytical standard, 10 mL

CAS Number:  
Price
3.790,00 kr
Log in for availability
1 * 10 mL
Product Details & Documents

For every lot exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.

Specifications

Specifications

  • Partikelstørrelse:
    20 µm
  • Pakn:
    10 mL
  • Standard:
    Analytical standard
  • Beskrivelse:
    Aqueous suspension, 10% solids
  • Nøjagtighed:
    std dev <0,3 µm
  • Cat. No.:
    74491-10ML-F
  • M-Clarity quality level:
    MQ100

Specifications