Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Geraniol, Supelco®

Geraniol, Supelco®

Glass bottle, 1 mL
Katalog # 48798-1ML
Irriterende
Leverandør:   Merck
Geraniol, Supelco®
Katalog # 48798-1ML
Leverandør:  Merck
CAS nummer:  106-24-1
Geraniol, Supelco®
Geraniol, Supelco®

Glass bottle, 1 mL

CAS nummer:  106-24-1
Price
1.290,00 kr
Log ind for tilgængelighed
1 * 1 mL
Product Details & Documents
Synonymer:  (E)-3,7-Dimethylocta-2,6-dien-1-ol
  • CAS nummer:106-24-1
  • Formel:C₁₀H₁₈O
  • MDL nummer:MFCD00002917
  • Molecular Weight:154.25 g/mol
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-377-1

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.