Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(±)-2-Butanol, Supelco®

(±)-2-Butanol, Supelco®

Glass bottle, 1 mL
Katalog # 96870-1ML-F
Brandfarlig
Irriterende
Leverandør:   Merck
(±)-2-Butanol, Supelco®
Katalog # 96870-1ML-F
Leverandør:  Merck
CAS nummer:  78-92-2
(±)-2-Butanol, Supelco®
(±)-2-Butanol, Supelco®

Glass bottle, 1 mL

CAS nummer:  78-92-2
Price
328,00 kr
Log ind for tilgængelighed
1 * 1 mL
Product Details & Documents
Synonymer:  (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
  • CAS nummer:78-92-2
  • Formel:H₃CCH₂CH(OH)CH₃
  • MDL nummer:MFCD00004569
  • Molecular Weight:74.12 g/mol
  • Merck Index:13,01540
  • Kogende Pt:94 °C (1013 hPa)
  • Smeltende Pt:–115 °C
  • Tæthed:0,8534 g/cm³ (20 °C)
  • Flash Pt:23 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:1120
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:201-158-5

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.