Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

(±)-2-Butanol, Supelco®

Glass bottle, 10 mL
Katalog # 96870-10ML-F
Brandfarlig
Irriterende
Leverandør:   Merck
(±)-2-Butanol, Supelco®
Katalog # 96870-10ML-F
Leverandør:  Merck
CAS nummer:  78-92-2
(±)-2-Butanol, Supelco®
(±)-2-Butanol, Supelco®

Glass bottle, 10 mL

CAS nummer:  78-92-2
Price
2.310,00 kr
Log ind for tilgængelighed
1 * 10 mL
Product Details & Documents
Synonymer:  (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
  • CAS nummer:78-92-2
  • Formel:H₃CCH₂CH(OH)CH₃
  • MDL nummer:MFCD00004569
  • Molecular Weight:74.12 g/mol
  • Merck Index:13,01540
  • Kogende Pt:94 °C (1013 hPa)
  • Smeltende Pt:–115 °C
  • Tæthed:0,8534 g/cm³ (20 °C)
  • Flash Pt:23 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:1120
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:201-158-5

 

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    10 mL

Specifikationer