Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
1-Propanol, Supelco®

1-Propanol, Supelco®

Glass bottle, 5 mL
Catalog # 96566-5ML-F
Flammable
Corrosive
Irritant
Supplier:   Merck
1-Propanol, Supelco®
Catalog # 96566-5ML-F
Supplier:  Merck
CAS Number:  71-23-8
1-Propanol, Supelco®
1-Propanol, Supelco®

Glass bottle, 5 mL

CAS Number:  71-23-8
Price
1.390,00 kr
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  Propylic alcohol, Propan-1-ol, n-Propan-1-ol, Ethylcarbinol, n-Propyl alcohol, Propanol-1, n-Propanol, Propyl alcohol, 1-Propyl alcohol, 1-Hydroxypropane
  • CAS Number:71-23-8
  • Formula:H₃CCH₂CH₂OH
  • MDL Number:MFCD00002941
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,07934
  • Boiling Pt:97 °C (1013 hPa)
  • Melting Pt:–127 °C
  • Density:0,8053 g/cm³ (20 °C)
  • Flash Pt:15 °C
  • Storage Temperature:Stue temperatur
  • UN:1274
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-746-9

Documents

Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    5 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.