Dichlormethan, Supelco®
Glass bottle, 500 mL
Katalog # SUPL66742-500ML
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Methylenchlorid
- CAS nummer:75-09-2
- Formel:CH₂Cl₂
- MDL nummer:MFCD00000881
- Molecular Weight:84.93 g/mol
- Merck Index:12,06140
- Kogende Pt:39,8 °C (1013 hPa)
- Smeltende Pt:–95 °C
- Tæthed:1,322 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-838-9
- FN:1593
- ADR:6.1,III
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



