Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
m-Cresol, Supelco®

m-Cresol, Supelco®

Glass bottle, 5 mL
Katalog # 65996-5ML-F
Giftig
Ætsende
Leverandør:   Merck
m-Cresol, Supelco®
Katalog # 65996-5ML-F
Leverandør:  Merck
CAS nummer:  108-39-4
m-Cresol, Supelco®
m-Cresol, Supelco®

Glass bottle, 5 mL

CAS nummer:  108-39-4
Price
865,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents
Synonymer:  m-Methylphenol
  • CAS nummer:108-39-4
  • Formel:CH₃C₆H₄OH
  • MDL nummer:MFCD00002302
  • Molecular Weight:108.14 g/mol
  • Merck Index:13,02604
  • Kogende Pt:202,2 °C (1013 hPa)
  • Smeltende Pt:11,5 °C
  • Tæthed:1,0336 g/cm³ (20 °C)
  • Flash Pt:73 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:2076
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-577-9
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    5 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.