Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
m-Cresol, Supelco®

m-Cresol, Supelco®

Glass bottle, 1 mL
Catalog # 65996-1ML-F
Toxic
Corrosive
Supplier:   Merck
m-Cresol, Supelco®
Catalog # 65996-1ML-F
Supplier:  Merck
CAS Number:  108-39-4
m-Cresol, Supelco®
m-Cresol, Supelco®

Glass bottle, 1 mL

CAS Number:  108-39-4
Price
295,00 kr
Log in for availability
1 * 1 mL
Product Details & Documents
Synonyms:  m-Methylphenol
  • CAS Number:108-39-4
  • Formula:CH₃C₆H₄OH
  • MDL Number:MFCD00002302
  • Molecular Weight:108.14 g/mol
  • Merck Index:13,02604
  • Boiling Pt:202,2 °C (1013 hPa)
  • Melting Pt:11,5 °C
  • Density:1,0336 g/cm³ (20 °C)
  • Flash Pt:73 °C
  • Storage Temperature:Stue temperatur
  • UN:2076
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-577-9
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    1 mL

Specifications