Supelco® Organic Reference Standard, (−)-Menthol
(−)-Menthol, Analytical standard, Glass bottle for solids, 1 g
Leverandør: Merck
Gå til:
Product Details & Documents
- GHS signal: GHS07
: P264 - P280 - P302 + P352 - P305 + P351 + P338 - P332 + P313 - P337 + P313
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g
- Standard:Analytical standard
- Beskrivelse:(−)-Menthol
- Cat. No.:63660-1G
- M-Clarity quality level:MQ100
- CAS-nr:2216-51-5
- Molekylvægt:156,27
- MDL No.:MFCD00062979
- Fareklassebeskrivelse:H315 - H319
- Flammepunkt:94 °C - closed cup
- Opbevaring betingelser:2...30 °C
- Melting point:~43 °C
- Synonym(s):(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol; 5-Methyl-2-(1-methylethyl)cyclohexanol
- Shipping conditions:STD



