Supelco® Organic Reference Standard, (−)-Menthol
(−)-Menthol, Analytical standard, Glass bottle for solids, 100 g
Katalog # 63660-100G
Leverandør: Merck
Gå til:
Product Details & Documents
- GHS signal: GHS07
Advarsel: P264 - P280 - P302 + P352 - P305 + P351 + P338 - P332 + P313 - P337 + P313
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g
- Standard:Analytical standard
- Beskrivelse:(−)-Menthol
- Cat. No.:63660-100G
- M-Clarity quality level:MQ100
- CAS-nr:2216-51-5
- Molekylvægt:156,27
- MDL No.:MFCD00062979
- Fareklassebeskrivelse:H315 - H319
- Flammepunkt:94 °C - closed cup
- Opbevaring betingelser:2...30 °C
- Melting point:~43 °C
- Synonym(s):(1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol; 5-Methyl-2-(1-methylethyl)cyclohexanol
- Shipping conditions:STD
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



