Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Nitrobenzen, Supelco®

Nitrobenzen, Supelco®

Glass bottle, 1 mL
Katalog # 06084-1ML-F
Giftig
Sundhed
Miljø
Leverandør:   Merck
Nitrobenzen, Supelco®
Katalog # 06084-1ML-F
Leverandør:  Merck
CAS nummer:  98-95-3
Nitrobenzen, Supelco®
Nitrobenzen, Supelco®

Glass bottle, 1 mL

CAS nummer:  98-95-3
Price
187,00 kr
Log ind for tilgængelighed
1 * 1 mL
Product Details & Documents
  • CAS nummer:98-95-3
  • Formel:C₆H₅NO₂
  • MDL nummer:MFCD00007043
  • Molecular Weight:123.11 g/mol
  • Merck Index:13,06621
  • Kogende Pt:210,8 °C (1013 hPa)
  • Smeltende Pt:5,7 °C
  • Tæthed:1,2037 g/cm³ (20 °C)
  • Flash Pt:88 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:1662
  • ADR:6.1,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:202-716-0

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.