Supelco® Organic Reference Standard, Alkane Standard Solution C₂₁-C₄₀
Alkane standard solution C₂₁-C₄₀, Analytical standard, 1 mL
Katalog # 04071-1ML
Leverandør: Merck
Supelco® Organic Reference Standard, Alkane Standard Solution C₂₁-C₄₀
Katalog # 04071-1ML
Leverandør: Merck
CAS nummer:
Gå til:
Product Details & Documents
- GHS signal: GHS02, GHS08, GHS07
Advarsel: P201 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331
Specifikationer
Specifikationer
- Pakn:1 mL
- Standard:Analytical standard
- Beskrivelse:Alkane standard solution C₂₁-C₄₀
- Cat. No.:04071-1ML
- M-Clarity quality level:MQ100
- Fareklassebeskrivelse:H225 - H304 - H315 - H336 - H361d - H373 - H412
- Flammepunkt:–4 °C - closed cup
- Opbevaring betingelser:2...30 °C
- Shipping conditions:STD



