Supelco® Organic Reference Standard, Dextran
Dextran, Analytical standard, for GPC, 270000, 500 mg
Katalog # 00894-500MG
Leverandør: Merck
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakn:500 mg
- Standard:Analytical standard, for GPC, 270000
- Beskrivelse:Dextran
- Cat. No.:00894-500MG
- M-Clarity quality level:MQ100
- CAS-nr:9004-54-0
- MDL No.:MFCD00130935
- Opbevaring betingelser:2...30 °C
- Shipping conditions:STD
- Formel:[C₆H₁₀O₅]ₙ
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



