Supelco® Organic Reference Standard, Dextran
Dextran, Analytical standard, for GPC, 270000, 100 mg
Katalog # 00894-100MG
Leverandør: Merck
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakn:100 mg
- Standard:Analytical standard, for GPC, 270000
- Beskrivelse:Dextran
- Cat. No.:00894-100MG
- M-Clarity quality level:MQ100
- CAS-nr:9004-54-0
- MDL No.:MFCD00130935
- Opbevaring betingelser:2...30 °C
- Shipping conditions:STD
- Formel:[C₆H₁₀O₅]ₙ



