Orders LinkContactUsLinkComponent
Capillaries for LC systems
Capillaries, non-swaged, male to female, 90 mm, I.Ø 0,12 mm, to connect column to DAD cell
Katalog # HEWLG1316-87314
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWLG1316-87314
Capillaries for LC systems
Capillaries for LC systems
Capillaries, non-swaged, male to female, 90 mm, I.Ø 0,12 mm, to connect column to DAD cell
Price
717,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
Specifikationer
  • Beskrivelse:
    Capillaries, non-swaged, male to female, 90 mm, I.Ø 0,12 mm, to connect column to DAD cell
  • Seq:
    37