Safety spectacles, A800
Anti-UV (5-2,5), anti-fog, grå, TSR-grå PC
Katalog # 111-0399
Leverandør: HONEYWELL SAFETY
Gå til:
Product Details & Documents
Single lens safety glasses for general protection against impact.
- Temples with soft and padded inserts with extra flex built in
- Slim profile for smaller faces
- Sporty styling provides a fashionable fit
Suitable for applications in utilities, manufacturing, biotech, laboratories, cleanrooms, ship building and automotive manufacturing.
Certifications: EN 166.
Specifikationer
- Model:Anti-UV (5-2,5), anti-fog
- Farve:grå
- Seq:04
- Linser:TSR-grå PC