Dækglas, Menzel-Gläser
Cover glasses, 40×60 mm, #1
Katalog # MENZBC040060A1
Leverandør: Epredia
Gå til:
Product Details & Documents
These cover slips are suitable for in vitro diagnostic (IVD) applications.
- Uniform quality of thickness
- No bubbles or flaws
- Made of pre-washed pure white Hydrolytic Class 1 glass
Specifikationer
- Antal:#1
- Beskrivelse:Cover glasses
- L×B:40×60 mm
- Seq:52